Learn More
Sino Biological Anti-ATP1B3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101309T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP1B3/CD298, and purified by antigen affinity chromatography.
Catalog No. 50-240-9038