Learn More
Sino Biological Recombinant Anti-ATP1B4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13027R108100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ATP1B4 (mature form, NP_036201.1; Tyr 132-Thr 357).
Catalog No. 50-255-4340