Learn More
Sino Biological Anti-ATP6V0A2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106994T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V0A2, and purified by antigen affinity chromatography.
Catalog No. 50-241-2523