Learn More
Sino Biological Anti-ATP6V0D1/VPATPD Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107633T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V0D1/VPATPD, and purified by antigen affinity chromatography.
Catalog No. 50-241-2444