Learn More
Sino Biological Anti-ATP6V1C1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106995T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ATP6V1C1, and purified by antigen affinity chromatography.
Catalog No. 50-241-2542