Learn More
Biomatik Corporation Human Atrophin 1 (ATN1) ELISA Kit
Supplier:  Biomatik Corporation EKU12540
Encompass_Preferred
Human Atrophin 1 (ATN1) ELISA Kit for detecting ATN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54259. Gene ID: 1822. Target Synonyms: B37; DRPLA; NOD; Dentatorubral Pallidoluysian Atrophy. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7949