Learn More
Raybiotech Inc C-Series Human Autophagy Array 1 Kit.
Catalog No. 501948485
Encompass_Preferred
Detects 20 Human Autophagic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ATG12, ATG7, ATG10, ATG13, ATG3, ATG4A, ATG4B, ATG5, Beclin, BNIP3L, DDR2, GABARAP, LAMP1 (CD107a), LC3a, LC3B, NBS1, P62, RHEB, MSK1, Alpha-synuclein
Catalog No. 50-194-8485 Supplier Raybiotech Inc Supplier No. AAHATG18
Detects 20 Human Autophagic Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ATG12, ATG7, ATG10, ATG13, ATG3, ATG4A, ATG4B, ATG5, Beclin, BNIP3L, DDR2, GABARAP, LAMP1 (CD107a), LC3a, LC3B, NBS1, P62, RHEB, MSK1, Alpha-synuclein