Learn More
Biomatik Corporation Human Autophagy Related Protein 5 (ATG5) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11079
Encompass_Preferred
Human Autophagy Related Protein 5 (ATG5) Mini sample ELISA Kit for detecting ATG5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H1Y0. Gene ID: 9474. Target Synonyms: ASP; APG5; APG5L; hAPG5; Apoptosis-specific protein; Autophagy protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6398