Learn More
Sino Biological Recombinant Human Autotaxin/ENPP-2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11308H07H50
Encompass_Preferred
A DNA sequence encoding the mature form of human ENPP2 (AAH34961.1) (Asp 49-Ile 863) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6974