Learn More
Biomatik Corporation Human Axis Inhibition Protein 2 (AXIN2) ELISA Kit
Supplier:  Biomatik Corporation EKU02654
Encompass_Preferred
Human Axis Inhibition Protein 2 (AXIN2) ELISA Kit for detecting AXIN2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2T1. Gene ID: 8313. Target Synonyms: Axil; Conductin; Axin Related Protein; Axin-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9107