Learn More
ACROBiosystems Biotinylated Human AXL Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502331504
Encompass_Preferred
Biotinylated Human AXL, Fc,Avitag (AXL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Trp 451 (Accession # P30530-1).
Catalog No. 50-233-1504 Supplier ACROBiosystems Supplier No. AXLH82F9200UG
Biotinylated Human AXL, Fc,Avitag (AXL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Trp 451 (Accession # P30530-1).