Learn More
Sino Biological Anti-AXL Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10279MM17TP25
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-0232