Learn More
Sino Biological Recombinant Human AXL Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 10279H05H100
Encompass_Preferred
A DNA sequence encoding the human AXL (AAH32229.1) (Met1-Pro449) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-224-6303