Learn More
ACROBiosystems ACROBiosystems Human Axl Protein, Fc Tag
Supplier:  ACROBiosystems AXLH52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human Axl, Fc Tag (AXL-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Trp 451 (Accession # P30530-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.6 kDa. The protein migrates as 95-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6347