Learn More
Sino Biological Anti-Zinc Alpha 2 Glycoprotein/AZGP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13242RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Alpha-2-glycoprotein / AZGP1 (rh Alpha-2-glycoprotein / AZGP1; Catalog#13242-H08H; P25311; Met1-Ser298). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4267