Learn More
Sino Biological Anti-Zinc Alpha 2 Glycoprotein/AZGP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204239T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Zinc Alpha 2 Glycoprotein/AZGP1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3892