Learn More
Sino Biological Anti-Zinc Alpha 2 Glycoprotein/AZGP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13242T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Zinc Alpha 2 Glycoprotein (rh Zinc Alpha 2 Glycoprotein; Catalog#13242-H08H; P25311; Met1-Ser298). Zinc Alpha 2 Glycoprotein specific IgG was purified by Human Zinc Alpha 2 Glycoprotein affinity chromatography.
Catalog No. 50-255-4265