Learn More
Sino Biological Anti-Azurocidin/CAP37 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10660RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AZU1 (rhAZU1; Catalog#10660-H08H; NP_001691.1; Met 1-Pro 250). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8174