Learn More
Apexbio Technology LLC Human B Cell Culture and Expansion Kit, 1 package
SDP
Supplier:  Apexbio Technology LLC BC10011
Encompass_Preferred
Human B cell culture and expansion kit is for activation and expansion of human B cells. Human monoclonal antibodies isolated for specific antibodies are widely used in immunity studies, historical studies and therapeutic development. Memory B cells derived from peripheral blood will produce high concentration of IgG in the supernatant under the stimulation of CD40L with other growth factors [1]. The B Cell Expansion Kit contains special treated feeder cells, expressing CD40L in activated form as well as human cell factors A and B potently inducing antibody-secret. The monoclonal antibody screening can be completed within 2 weeks with the help of the kit. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8465