Learn More
STEMCELL Technologies ImmunoCult™ Human B Cell Expansion Kit
SDP
Catalog No. NC1988309
Encompass
Reliably expand human B cells that are ready for use in downstream assays and applications—without the use of serum, specialized cultureware, or problematic feeder cells.ImmunoCult™ Human B Cell Expansion Kit comprises ImmunoCult™-ACF Human B Cell Expansion Supplement and ImmunoCult™-XF B Cell Base Medium, which together ensure consistent activation and expansion of human B cells and their maturation to plasma cells. B cells can be harvested and used directly in downstream applications after just one week or less in culture. For your convenience, all kit components are also available for individual purchase.This kit is compatible with many of our other upstream and downstream products. For example, you can use EasySep™ cell separation kits to isolate a range of B cell subsets—such as pan-B cells, memory B cells, and naïve B cells—that can be isolated and then immediately expanded using ImmunoCult™ Human B Cell Expansion Kit.
Catalog No. NC1988309 Supplier STEMCELL Technologies Supplier No. 1000645
Reliably expand human B cells that are ready for use in downstream assays and applications—without the use of serum, specialized cultureware, or problematic feeder cells.ImmunoCult™ Human B Cell Expansion Kit comprises ImmunoCult™-ACF Human B Cell Expansion Supplement and ImmunoCult™-XF B Cell Base Medium, which together ensure consistent activation and expansion of human B cells and their maturation to plasma cells. B cells can be harvested and used directly in downstream applications after just one week or less in culture. For your convenience, all kit components are also available for individual purchase.This kit is compatible with many of our other upstream and downstream products. For example, you can use EasySep™ cell separation kits to isolate a range of B cell subsets—such as pan-B cells, memory B cells, and naïve B cells—that can be isolated and then immediately expanded using ImmunoCult™ Human B Cell Expansion Kit.