Learn More
American Research Products Inc Human B cell leukemia lymphoma
Catalog No. 5000329551
Encompass
Human B-cell leukemia/lymphoma 2 Bcl-2 ELISA Kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 225 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29551 Supplier American Research Products Inc Supplier No. CSBE08853H
Human B-cell leukemia/lymphoma 2 Bcl-2 ELISA Kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 225 ng/mL Sample serum plasma tissue homogenates Method Sandwich