Learn More
American Research Products Inc Human b galactosidase bGAL EL
Catalog No. 5000328696
Encompass
Human b-galactosidase bGAL ELISA Kit from CUSABIO Detection Range 7 8 lU/mL-500 lU/mL Sensitivity 1 95 lU/mL Sample serum plasma urine tissue homogenates Method Sandwich
Catalog No. 50-003-28696 Supplier American Research Products Inc Supplier No. CSBE09463H
Human b-galactosidase bGAL ELISA Kit from CUSABIO Detection Range 7 8 lU/mL-500 lU/mL Sensitivity 1 95 lU/mL Sample serum plasma urine tissue homogenates Method Sandwich