Learn More
Sino Biological Anti-Beta-2 microglobulin Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11976MM35P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Beta-2 microglobulin/B2M (rh Beta-2 microglobulin/B2M; Catalog#11976-H08H; NP_004039.1; Met1-Met119) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8689