Learn More
Sino Biological Anti-Beta-2 microglobulin Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 11976MM34H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Beta-2 microglobulin/B2M (rh Beta-2 microglobulin/B2M; Catalog#11976-H08H; NP_004039.1; Met1-Met119). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1108