Learn More
Sino Biological Recombinant Anti-Beta-2 microglobulin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11976R209100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human B2M / beta-2 microglobulin (rh B2M / beta-2 microglobulin; Catalog#11976-H08H; NP_004039.1; Met1-Met119).
Catalog No. 50-241-1993