Learn More
Sino Biological Recombinant Human CD57/B3gat1 Protein (mFc Tag)
Supplier:  Sino Biological 18529H04H200
Encompass_Preferred
A DNA sequence encoding the human B3GAT1 (NP_473366.1) (Thr28-Ile334) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-255-4001