Learn More
Sino Biological Anti-B3GAT3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12290T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B3GAT3 / GLCATI (rh B3GAT3 / GLCATI; Catalog#12290-H07E; NP_036332.2; Glu72-Val335). B3GAT3 / GLCATI specific IgG was purified by Human B3GAT3 / GLCATI affinity chromatography.
Catalog No. 50-241-2533