Learn More
Raybiotech Inc Human B4GalT1 ELISA Kit
Supplier:  Raybiotech Inc ELHB4GALT11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.32 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human B4GalT1.
Catalog No. 50-194-9008