Learn More
Sino Biological Recombinant Anti-B4GALT1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11220R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human B4GALT1 / GGTB2 (rh B4GALT1; Catalog#11220-H07H; Gly 44-Ser 398; NP_001488.2).
Catalog No. 50-255-4778