Learn More
Sino Biological Anti-B4GALT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11220RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human B4GALT1 / GGTB2 (rh B4GALT1; Catalog#11220-H07H; NP_001488.2; Gly 44-Ser 398).Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4777