Learn More
Bon Opus Biosciences Recombinant Human CD275/B7-H2 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP100350UG
Encompass_Preferred
Recombinant Human CD275/B7-H2 (C-mFc) is produced by Human Cells. The target gene encoding Asp19-Thr256 is expressed with a C-mFc tag. Accession# O75144. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1003
Catalog No. 50-217-2605