Learn More
Raybiotech Inc Human B7-H2/ICOS Ligand ELISA Kit
Supplier:  Raybiotech Inc ELHB7H21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human B7-H2.
Catalog No. 50-194-9010