Learn More
ACROBiosystems PE-Labeled Human B7-H3 (4Ig), His Tag (B7B-HP2H9) is produced via site-specific conjugation of PE to Human B7-H3 (4Ig), His Tag under optimal conditions with a proprietary technology. It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1).
Catalog No. 502231434
Encompass_Preferred
Human B7 homolog 3 (B7-H3) is a member of the B7 family of immune proteins that provide signals for the regulation of immune responses. Other family members include B7-1, B7-2, B7-H1/PD-L1, B7-H2, and PD-L2. B7 family proteins are type I transmembrane immunoglobulin (Ig) superfamily members that contain extracellular Ig V‑like and Ig C‑like domains with a short cytoplasmic tail. Termed 4IgB7-H3 or B7-H3b, this molecule has two additional Ig-like domains (one V‑type and one C‑type) and shows a ubiquituous expression pattern.
Catalog No. 50-223-1434 Supplier ACROBiosystems Supplier No. B7BHP2H925TESTS
Human B7 homolog 3 (B7-H3) is a member of the B7 family of immune proteins that provide signals for the regulation of immune responses. Other family members include B7-1, B7-2, B7-H1/PD-L1, B7-H2, and PD-L2. B7 family proteins are type I transmembrane immunoglobulin (Ig) superfamily members that contain extracellular Ig V‑like and Ig C‑like domains with a short cytoplasmic tail. Termed 4IgB7-H3 or B7-H3b, this molecule has two additional Ig-like domains (one V‑type and one C‑type) and shows a ubiquituous expression pattern.