Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human B7-H3 (CD276) (4Ig) Fc Recombinant Protein, PeproTech®
GREENER_CHOICE

Catalog No. 5000331930
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-003-31930 500 μg
50-003-31919 100 μg
31-050-10UG 10 μg
31-050-1MG 1 mg
50-003-31929 250 μg
31-050-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50-003-31930 Supplier Gibco™ Supplier No. 31050500UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human B7-H3 (4Ig) Fc is a 145.0 kDa glycosylated, disulfide-linked homodimer of 666 amino acid residues. This product is shipped at ambient temperature. For storage, handling and resonstitution information, please see the lot-specif Certificate of Analysis.

CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5ZPR3
For Use With (Application) Binding Assay, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 80381
Molecular Weight (g/mol) 72.5 kDa
Name Human B7-H3 (CD276) (4Ig) Fc
Purification Method Purified
Quantity 500 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352
Common Name B7-H3 (CD276)
Gene Symbol CD276
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence LEVQVPEDPV VALVGTDATL CCSFSPEPGF SLAQLNLIWQ LTDTKQLVHS FAEGQDQGSA YANRTALFPD LLAQGNASLR LQRVRVADEG SFTCFVSIRD FGSAAVSLQV AAPYSKPSMT LEPNKDLRPG DTVTITCSSY QGYPEAEVFW QDGQGVPLTG NVTTSQMANE QGLFDVHSIL RVVLGANGTY SCLVRNPVLQ QDAHSSVTIT PQRSPTGAVE VQVPEDPVVA LVGTDATLRC SFSPEPGFSL AQLNLIWQLT DTKQLVHSFT EGRDQGSAYA NRTALFPDLL AQGNASLRLQ RVRVADEGSF TCFVSIRDFG SAAVSLQVAA PYSKPSMTLE PNKDLRPGDT VTITCSSYRG YPEAEVFWQD GQGVPLTGNV TTSQMANEQG LFDVHSVLRV VLGANGTYSC LVRNPVLQQD AHGSVTITGQ PMTGGPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK
Activity Determined by its ability to bind human B7-H3 antibody in a functional ELISA.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.