Learn More
Bon Opus Biosciences Recombinant Human B7H3-4Ig/B7-H3b (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP100950UG
Encompass_Preferred
Recombinant Human B7H3-4Ig/B7-H3b (C-hFc) is produced by Human Cells. The target gene encoding Gly27-Thr461 is expressed with a C-hFc tag. Accession# Q5ZPR3. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1009
Catalog No. 50-217-2611