Learn More
Bon Opus Biosciences Recombinant Human B7H3-4Ig/B7-H3b (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP101050UG
Encompass_Preferred
Recombinant Human B7H3-4Ig/B7-H3b (C-mFc) is produced by Human Cells. The target gene encoding Gly27-Thr461 is expressed with a C-mFc tag. Accession# Q5ZPR3. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1010
Catalog No. 50-217-2612