Learn More
ACROBiosystems ACROBiosystems Human B7-H5 / Gi24 / VISTA Protein, His Tag (MALS verified)
Catalog No. 502106388
Encompass_Preferred
Recombinant Protein;100UG;Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.0 kDa. The protein migrates as 33-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human VSIG3, Fc Tag (Cat. No. VS3-H5258) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human B7-H5, His Tag (Cat. No. B75-H52H0) with an affinity constant of 53.4 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-210-6388 Supplier ACROBiosystems Supplier No. B75H52H0100UG
Recombinant Protein;100UG;Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.0 kDa. The protein migrates as 33-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Human VSIG3, Fc Tag (Cat. No. VS3-H5258) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human B7-H5, His Tag (Cat. No. B75-H52H0) with an affinity constant of 53.4 μM as determined in a SPR assay (Biacore T200) (Routinely tested).