Learn More
Bon Opus Biosciences Recombinant Human B7-H7 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP101350UG
Encompass_Preferred
Recombinant Human B7-H7 (C-mFc) is produced by Human Cells. The target gene encoding Ile23-Asn344 is expressed with a C-mFc tag. Accession# Q9UM44. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1013
Catalog No. 50-217-2615