Learn More
Raybiotech Inc Human B7-H1 (CD274) ELISA Kit
Supplier:  Raybiotech Inc ELHB7H11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 46 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human B7-H1 (CD274)
Catalog No. 50-194-9009