Learn More
Sino Biological Recombinant Human B7-H7/HHLA2 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 16139H08HB20
Encompass_Preferred
A DNA sequence encoding the human HHLA2 (NP_009003.1) (Met1-Asn344) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-224-7224