Learn More
ACROBiosystems ACROBiosystems Human BACE-1 Protein, Tag Free (active enzyme, MALS verified)
Supplier:  ACROBiosystems BA1H5213100UG
Encompass_Preferred
Recombinant Protein;100UG;Human BACE-1, Tag Free (BA1-H5213) is expressed from human 293 cells (HEK293). It contains AA Thr 22 - Thr 457 (Accession # NP_036236.1).;This protein carries no "tag". The protein has a calculated MW of 49.0 kDa. The protein migrates as 53-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6403