Learn More
Biotang Inc Human BACE1 / ASP2 Protein, 50ug, The mature form of human BACE1 (NP_036236.1) extracellular domain (Met1-Thr 457) with a quinary-aa peptide (DDDDK) at the C-terminus was expressed in human cells and purified.
Supplier:  Biotang Inc RPR9917
Encompass_Preferred
Human BACE1 / ASP2 Protein, 50ug, The mature form of human BACE1 (NP_036236.1) extracellular domain (Met1-Thr 457) with a quinary-aa peptide (DDDDK) at the C-terminus was expressed in human cells and purified.
Catalog No. 50-751-5342