Learn More
Sino Biological Human BACE1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10064H02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 457) of human BACE1 (NP_036236.1) was expressed as a secreted chimeric protein with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-237-2368