Learn More
Biomatik Corporation Human Baculoviral IAP Repeat Containing Protein 6 (BIRC6) ELISA Kit
Supplier:  Biomatik Corporation EKU02659
Encompass_Preferred
Human Baculoviral IAP Repeat Containing Protein 6 (BIRC6) ELISA Kit for detecting BIRC6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NR09. Gene ID: 57448. Target Synonyms: BRUCE; Apollon; BIR repeat-containing ubiquitin-conjugating enzyme; Ubiquitin-conjugating BIR domain enzyme apollon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4887