Learn More
ACROBiosystems Human BAFFR 25tests
Supplier:  ACROBiosystems BARHP2H625TESTS
Encompass_Preferred
PE-Labeled Human BAFFR Protein His Tag (BAR-HP2H6) is produced via site-specific conjugation of PE to Human BAFFR Protein His Tag under optimal conditions with a proprietary technology. Human BAFFR Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).
Catalog No. 50-253-5478