Learn More
ACROBiosystems Human BAFFR / TNFRSF13C Protein, His Tag
Catalog No. 502055223
Encompass_Preferred
Recombinant Protein;1MG;Human BAFFR Protein, His Tag (BAR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.6 kDa. The protein migrates as 13-20 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Human BAFFR Protein, His Tag (Cat. No. BAR-H52H3) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human BAFFR Protein, His Tag (Cat. No. BAR-H52H3) at 2 μg/mL (100 μL/well) can bind Anti-BAFFR Antibody, Human IgG1 with a linear range of 0.1-5 ng/mL (Routinely tested).
Catalog No. 50-205-5223 Supplier ACROBiosystems Supplier No. BARH52H31MG
Recombinant Protein;1MG;Human BAFFR Protein, His Tag (BAR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.6 kDa. The protein migrates as 13-20 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 1 μg/mL (100 μL/well) can bind Human BAFFR Protein, His Tag (Cat. No. BAR-H52H3) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human BAFFR Protein, His Tag (Cat. No. BAR-H52H3) at 2 μg/mL (100 μL/well) can bind Anti-BAFFR Antibody, Human IgG1 with a linear range of 0.1-5 ng/mL (Routinely tested).