Learn More
BIOHIPPO Human BALP Microsample
SDP
Catalog No. 5000314340
Encompass
Human BALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BALP in serum plasma and other biological fluids It provides a detection range of 1 25-80 ng/mL with a sensitivity of 0 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14340 Supplier BIOHIPPO Supplier No. BHE15210684
Human BALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BALP in serum plasma and other biological fluids It provides a detection range of 1 25-80 ng/mL with a sensitivity of 0 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color