Learn More
Sino Biological Human BAMBI Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785225
Encompass_Preferred
The human BAMBI / NMA ELISA Pair Set is for the quantitative determinationof human BAMBI / NMA.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for BAMBI / NMA coated on a 96-well plate. Standards and samples are added to the wells, and any BAMBI / NMA present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-BAMBI / NMA monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of BAMBI / NMA present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85225 Supplier Sino Biological Supplier No. SEK1089015
The human BAMBI / NMA ELISA Pair Set is for the quantitative determinationof human BAMBI / NMA.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for BAMBI / NMA coated on a 96-well plate. Standards and samples are added to the wells, and any BAMBI / NMA present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-BAMBI / NMA monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of BAMBI / NMA present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.