Learn More
Sino Biological Human BAMBI PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50403R105100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BAMBI (rh BAMBI; Catalog#10890-H08H; NP_036474.1; Met 1-Ala 152). BAMBI specific IgG was purified by human BAMBI affinity chromatography.
Catalog No. 50-237-4087