Learn More
Biomatik Corporation Human Basic Salivary Proline Rich Protein 2 (PRB2) ELISA Kit
Supplier:  Biomatik Corporation EKU02663
Encompass_Preferred
Human Basic Salivary Proline Rich Protein 2 (PRB2) ELISA Kit for detecting PRB2 in saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.52ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02812. Gene ID: 653247. Target Synonyms: PRPPRB1; Ps; cP7; Proline-Rich Protein BstNI Subfamily 2; Con1 glycoprotein; Basic peptide P-F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9105